1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Butyl-3-methylpyridin-1-ium tetrafluoroborate
H26983.18
H26983.06
H26983.18EA
257
EUR
H26983.18
H26983.06
1-Butyl-3-methylpyridiniumtetrafluoroborate ≥99%
1-Butyl-3-methylpyridinium tetrafluoroborate
Purity: 99%
CAS No.: 597581-48-1
Molecular Formula: C10H16BF4N
Formula Weight: 237.06
MDL No.: MFCD08458499
Formula:
C₁₀H₁₆BF₄N MW: 237,05 g/mol Density: 1,204 Storage Temperature: Ambient |
MDL Number:
MFCD08458499 CAS Number: 597581-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...