1-Butyl-4-methylpyridiniumtetrafluoroborate ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Butyl-4-methylpyridin-1-ium tetrafluoroborate

H27100.06 H27100.18
H27100.06EA 94.1 EUR
H27100.06 H27100.18
1-Butyl-4-methylpyridiniumtetrafluoroborate ≥99%
1-Butyl-4-methylpyridinium tetrafluoroborate

Purity: 99% CAS No.: 343952-33-0 Molecular Formula: C10H16BF4N Formula Weight: 237.06 Refractive Index: 1.4

Formula: C₁₀H₁₆BF₄N
MW: 237,05 g/mol
Density: 1,2
Storage Temperature: Ambient
MDL Number: MFCD03095460
CAS Number: 343952-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR