4-Bromo-5-fluoro-2-nitrotoluene ≥98%

Supplier: Thermo Scientific
Danger

H27552.06 H27552.03
H27552.06EA 95 EUR
H27552.06 H27552.03
4-Bromo-5-fluoro-2-nitrotoluene ≥98%
4-Bromo-5-fluoro-2-nitrotoluene

Purity: 98% CAS No.: 224185-19-7 Molecular Formula: C7H5BrFNO2 Formula Weight: 234.02 Melting Point: 59-63° MDL No.: MFCD04039299

Formula: C₇H₅BrFNO₂
MW: 234,02 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD04039299
CAS Number: 224185-19-7
EINECS: 262-536-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR