4-(Boc-amino)-3-iodopyridine 96%

Supplier: Thermo Scientific
Warning

Synonyms: N-Boc-4-Amino-3-iodopyridine

H28751.03
H28751.03EA 150 EUR
H28751.03
4-(Boc-amino)-3-iodopyridine 96%
4-(Boc-amino)-3-iodopyridine

Purity: 96% CAS No.: 211029-67-3 Molecular Formula: C10H13IN2O2 Formula Weight: 320.13 Melting Point: 79-83° MDL No.: MFCD04973409

Formula: C₁₀H₁₃IN₂O₂
MW: 320,13 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD04973409
CAS Number: 211029-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR