1-(2-Aminoethyl)-4-methylpiperazine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-(4-Methylpiperazin-1-yl)ethanamine, 2-(4-Methyl-piperazin-1-yl)-ethylamine

H52371.MD H52371.03 H52371.06
H52371.MDEA 74.6 EUR
H52371.MD H52371.03 H52371.06
1-(2-Aminoethyl)-4-methylpiperazine ≥97%
1-(2-Aminoethyl)-4-methylpiperazine

Purity: 97+% CAS No.: 934-98-5 Molecular Formula: C7H17N3 Formula Weight: 143.43 UN No.: 2735 MDL No.: MFCD03701701

Formula: C₇H₁₇N₃
MW: 143.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701701
CAS Number: 934-98-5
EINECS: 213-296-3
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR