2,6-Dichloro-3-nitrobenzonitrile ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,6-Dichloro-3-nitrobenzenecarbonitrile

L06186.03 L06186.06
L06186.03EA 36.4 EUR
L06186.03 L06186.06
2,6-Dichloro-3-nitrobenzonitrile ≥98%
2,6-Dichloro-3-nitrobenzonitrile

Purity: 98% CAS No.: 5866-98-8 Molecular Formula: C7H2Cl2N2O2 Formula Weight: 217.01 Melting Point: 106-109° Beilstein Reference: 2113045 UN No.: 3439 MDL No.: MFCD00051513

Formula: C₇H₂Cl₂N₂O₂
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00051513
CAS Number: 5866-98-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR