3-Furoic acid ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: Furan-3-carboxylic acid, 3-Furancarboxylic acid

L11629.06 L11629.14
L11629.06EA 50.4 EUR
L11629.06 L11629.14
3-Furoic acid ≥99%
3-Furoic acid

Purity: 99% CAS No.: 488-93-7 Molecular Formula: C5H4O3 Formula Weight: 112.08 Melting Point: 120-123° Beilstein Reference: 108638

Formula: C₅H₄O₃
MW: 112,08 g/mol
Melting Pt: 120…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00005350
CAS Number: 488-93-7
EINECS: 207-689-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR