2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate ≥95%
Supplier: Thermo Fisher Scientific
L16304.03
L16304.06
L16304.03EA
0
EUR
L16304.03
L16304.06
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate ≥95%
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate
Purity: 95%
CAS No.: 59643-43-5
Molecular Formula: C15H23F3O4S
Formula Weight: 356.41
Melting Point: ca 163° dec.
Beilstein Reference: 3644038
Formula:
C₁₅H₂₃F₃O₄S MW: 356.41 g/mol Melting Pt: ∼163 °C (dec.) |
MDL Number:
MFCD00229854 CAS Number: 59643-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...