Brij® 35

Supplier: G Biosciences
Warning

Synonyms: Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23

786-521 786-351
786-521EA 126 EUR
786-521 786-351
Brij® 35
Brij® 35
Formula: C₅₈H₁₁₈O₂₄
MW: 1199,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080891
CAS Number: 9002-92-0
EINECS: 500-002-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR