Diisodecyl phthalate, Sigma-Aldrich®

Supplier: Merck

Synonyms: DIDP, Phthalic acid diisodecyl ester

M-Clarity™ quality level = MQ200

8.14732.0100 8.14732.1000
8.14732.0100EA 42.7 EUR
8.14732.0100 8.14732.1000
Diisodecyl phthalate, Sigma-Aldrich®
Diisodecyl phthalate
Formula: C₂₈H₄₆O₄
MW: 446,67 g/mol
MDL Number: MFCD00048389
CAS Number: 26761-40-0
EINECS: 247-977-1

Order Now

Specification Test Results

Solubility 0.00028 g/l (24 °C)
Melting Point -50 °C
Molar Mass 446.66 g/mol
Boiling Point 250 - 267 °C (7 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 240 °C
Density 0.97 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 380 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR