TES sodium salt (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt)
Supplier: Roth Carl
6976.1
6976.2
ROTH6976.1EA
0
EUR
ROTH6976.1
ROTH6976.2
TES sodium salt (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt)
TES sodium salt (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid sodium salt)
Formula:
C₆H₁₄NNaO₆S Storage Temperature: Ambient |
MDL Number:
MFCD00065482 CAS Number: 70331-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...