Diisodecyl phthalate, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

80135-10ML 80135-50ML
80135-10MLEA 59.6 EUR
80135-10ML 80135-50ML
Diisodecyl phthalate, Supelco®
Diisodecyl phthalate
Formula: C₂₈H₄₆O₄
MDL Number: MFCD00048389
CAS Number: 26761-40-0
EINECS: 247-977-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b1b19fbba6f469fa073ae4f64ce8de05bcd01b6&vid=3e0c742e35db6f7b1fcbd681ca79c4d44d788e97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR