1-Ethynylcyclopentanol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98+%
CAS No.: 17356-19-3
Molecular Formula: C7H10O
Formula Weight: 110.16
Flash Point: 48°(118°F)
Refractive Index: 1.4740
Odour: Irritating
Melting Point: 23-27°
Boiling Point: 156-159°
Beilstein Reference: 1924167
UN No.: 1325
MDL No.: MFCD00001365
Formula:
C₇H₁₀O Boiling Pt: 156…159 °C Melting Pt: 23…27 °C Density: 0,962 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003858 CAS Number: 17356-19-3 EINECS: 241-385-7 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...