Cyclohexylacetone 96%

Supplier: Thermo Fisher Scientific

A12748.06 A12748.14
A12748.06EA 200 EUR
A12748.06 A12748.14
Cyclohexylacetone 96%
Cyclohexylacetone

Refractive Index: 1.4528
Boiling Point: 198-200°
MDL No.: MFCD00019395

Formula: C₉H₁₆O
Boiling Pt: 198…200 °C
Density: 0,905
Storage Temperature: Ambient
MDL Number: MFCD00019396
CAS Number: 103-78-6
EINECS: 203-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR