2,4,4-Trimethylcyclopentanone ≥98%

Supplier: Thermo Fisher Scientific
Warning

B20425.03 B20425.06 B20425.14
B20425.03EA 40 EUR
B20425.03 B20425.06 B20425.14
2,4,4-Trimethylcyclopentanone ≥98%
2,4,4-Trimethylcyclopentanone

Purity: 98% CAS No.: 4694-12-6 Molecular Formula: C8H14O Formula Weight: 126.20 Flash Point: 43°(110°F) Refractive Index: 1.4320 Boiling Point: 159-161° UN No.: 1224 MDL No.: MFCD00019313

Formula: C₈H₁₄O
Boiling Pt: 159…161 °C
Melting Pt: –25,6 °C
Density: 0,877
Flash Pt: 43 °C (110 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019313
CAS Number: 4694-12-6
EINECS: 225-155-3
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR