Methylenecyclohexane 98%
Supplier: Thermo Scientific
|
Danger
|
325510010
325510050
ACRO325510010EA
56
EUR
ACRO325510010
ACRO325510050
Methylenecyclohexane 98%
Methylenecyclohexane
Formula:
C₇H₁₂ Boiling Pt: 102…103 °C (760 mmHg) Melting Pt: –126 °C Density: 0,8 g/cm³ Flash Pt: –006 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001661 CAS Number: 1192-37-6 EINECS: 203-624-3 UN: 1993 ADR: 3,II Merck Index: 13,06074 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4465 to 1.4505 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...