Cycloheptene 96% Stabilised

Supplier: Thermo Fisher Scientific
Danger


Stabilisation: BHT

H29062.14 H29062.06
H29062.14EA 373 EUR
H29062.14 H29062.06
Cycloheptene 96% Stabilised
Cycloheptene

Stabilized with 0.1% BHT


Purity: 96% CAS No.: 628-92-2 Molecular Formula: C7H12 Formula Weight: 96.17 Flash Point: -7°(19°F) Refractive Index: 1.458 Boiling Point: 112-115° UN No.: 2242 MDL No.: MFCD00004156

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 112…115 °C
Melting Pt: –56 °C
Density: 0.833
Flash Pt: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR