Cyclopentene 95+%

Supplier: Thermo Scientific
Danger

295905000 295900025 295901000
ACRO295905000EA 178 EUR
ACRO295905000 ACRO295900025 ACRO295901000
Cyclopentene 95+%
Cyclopentene
Formula: C₅H₈
Boiling Pt: 44 °C (760 mmHg)
Melting Pt: –135 °C
Density: 0,774 g/cm³
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001394
CAS Number: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20°C, 589 nm)
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR