Cyclopentene ≥97%

Supplier: Thermo Fisher Scientific
Danger

A11027.AE A11027.AP
A11027.AEEA 49.9 EUR
A11027.AE A11027.AP
Cyclopentene ≥97%
Cyclopentene

Purity: 97% CAS No.: 142-29-0 Molecular Formula: C5H8 Formula Weight: 68.12 Storage Temperature: 2°C to 8°C Flash Point: -29°(-20°F) Refractive Index: 1.4225 Melting Point: -135° Boiling Point: 44-45° Beilstein Reference: 635707 UN No.: 2246 MDL No.: MFCD00001394

Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 44…45 °C
Melting Pt: –135 °C
Density: 0.773
Flash Pt: –29 °C (–20 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001394
CAS Number: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR