Potassium bromide
Supplier: Roth Carl
|
Warning
|
A137.1
A137.2
ROTHA137.1EA
34.7
EUR
ROTHA137.1
ROTHA137.2
Potassium bromide
Potassium bromide
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 Melting Pt: 730 Density: 2.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...