Sodium bromate ≥99.5% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
14101.22
14101.36
14101.A3
14101.22EA
28.7
EUR
14101.22
14101.36
14101.A3
Sodium bromate ≥99.5% (metals basis)
Sodium bromate
Purity: 99.5%
CAS No.: 7789-38-0
Molecular Formula: NaBrO3
Formula Weight: 150.90
Physical Form: Crystalline
Density: 3.34
Refractive Index: 1.594
Appearance: White
Odour: Odourless
Melting Point: 381° dec.
Solubility: Soluble in water (2.5 parts in cold water, 1.1 part in boiling water)
Merck Reference: 14,8593
UN No.: 1494
MDL No.: MFCD00003476
Formula:
BrNaO₃ MW: 150,89 g/mol Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 381 °C (dec.) Density: 3,34 Storage Temperature: Ambient |
MDL Number:
MFCD00003476 CAS Number: 7789-38-0 EINECS: 232-160-4 UN: 1494 ADR: 5.1,II Merck Index: 14,8593 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...