Rubidium chloride 99+%

Supplier: Thermo Fisher Scientific

Synonyms: Rubidiumchloride

201270250 201271000
ACRO201270250EA 257 EUR
ACRO201270250 ACRO201271000
Rubidium chloride 99+%
Rubidium chloride
Formula: RbCl
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 718 °C
Density: 2,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total trace metal impurities ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR