Lithium amide 95%
Supplier: Thermo Scientific
|
Danger
|
L16023.14
L16023.22
L16023.36
L16023.14EA
35.6
EUR
L16023.14
L16023.22
L16023.36
Lithium amide 95%
Lithium amide
Purity: 95%
CAS No.: 7782-89-0
Molecular Formula: H2LiN
Formula Weight: 22.96
Melting Point: 373°
Boiling Point: 430°
Merck Reference: 14,5522
UN No.: 1390
MDL No.: MFCD00011093
Formula:
H₂LiN MW: 22,96 g/mol Boiling Pt: 430 °C Melting Pt: 373 °C Density: 1,178 Storage Temperature: Ambient |
MDL Number:
MFCD00011093 CAS Number: 7782-89-0 EINECS: 231-968-4 UN: 1390 ADR: 4.3,II Merck Index: 14,5522 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...