Sodium amide 95%, pure
Supplier: Thermo Scientific
|
Danger
|
197051000
197055000
197050025
ACRO197051000EA
50.2
EUR
ACRO197051000
ACRO197055000
ACRO197050025
Sodium amide 95%, pure
Sodium amide
Formula:
H₂NNa MW: 39,01 g/mol Boiling Pt: 400 °C Melting Pt: 210 °C Density: 1,39 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011117 CAS Number: 7782-92-5 EINECS: 231-971-0 UN: 3131 ADR: 4.3,I Merck Index: 12,08717 |
Specification Test Results
Appearance | White to greyish-white Powder or irregular granules or flakes |
Titration with HClO4 | ≥94 % |
Free Sodium | Passes test (A small amount dropped into water, does not catch fire) |
Peroxides | ≤0.1 % (as NaNH2O2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...