Potassium nitrite 96+% ACS
Supplier: Thermo Fisher Scientific
|
Danger
|
423060050
423065000
423060025
ACRO423060050EA
26
EUR
ACRO423060050
ACRO423065000
ACRO423060025
Potassium nitrite 96+% ACS
Potassium nitrite
Formula:
KNO₂ Melting Pt: 387 °C Density: 1,92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011408 CAS Number: 7758-09-0 EINECS: 231-832-4 UN: 1488 ADR: 5.1,II Merck Index: 13,07734 |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Titration with KMnO4 | ≥96.0 % |
Chloride (Cl) | ≤300 ppm |
Sulfate (SO4) | ≤100 ppm |
Calcium (Ca) | ≤50 ppm |
Iron (Fe) | ≤10 ppm |
Magnesium (Mg) | ≤20 ppm |
Sodium (Na) | ≤5000 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...