Lithium peroxide ≥95%
Supplier: Thermo Fisher Scientific
|
Danger
|
13421.22
13421.36
13421.22EA
517
EUR
13421.22
13421.36
Lithium peroxide ≥95%
Lithium peroxide
Purity: 95%
CAS No.: 12031-80-0
Molecular Formula: Li2O2
Formula Weight: 45.88
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White to off-white
Odour: Odourless
UN No.: 1472
MDL No.: MFCD00016184
Formula:
Li₂O₂ MW: 45.88 g/mol Density: 1.2063 g/cm³ (18 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016184 CAS Number: 12031-80-0 EINECS: 234-758-0 UN: 1472 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...