Dipotassium hexachlororhenate ≥99.9% (metals basis)

Supplier: Thermo Scientific
Warning

Synonyms: Potassium hexachlororhenate(IV)

62113.03 62113.06
62113.03EA 153 EUR
62113.03 62113.06
Dipotassium hexachlororhenate ≥99.9% (metals basis)
di-Potassium hexachlororhenate

Purity: 99.9% CAS No.: 16940-97-9 Molecular Formula: Cl6K2Re Formula Weight: 477.12 Physical Form: Crystalline Density: 3.34 Appearance: Yellow to green Odour: Odourless MDL No.: MFCD00058746

Formula: Cl₆K₂Re
MW: 477,12 g/mol
Density: 3,34
Storage Temperature: Ambient
MDL Number: MFCD00058746
CAS Number: 16940-97-9
EINECS: 241-008-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR