Sulphuric acid 96,00% (+/- 0,15%) 96.00% (±0.15%) water, pure

Supplier: Thermo Fisher Scientific
Danger

302070010 302070025 302070011 302070026 302070100
ACRO302070010EA 63 EUR
ACRO302070010 ACRO302070025 ACRO302070011 ACRO302070026 ACRO302070100
Sulphuric acid 96,00% (+/- 0,15%) 96.00% (±0.15%) water, pure
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Titration with NaOH 95.85 to 96.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR