Selenious acid 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Selenous acid
198870500
198872500
198870010
ACRO198870500EA
86.3
EUR
ACRO198870500
ACRO198872500
ACRO198870010
Selenious acid 98%
Selenious acid
Formula:
H₂O₃Se MW: 128,97 g/mol Boiling Pt: 315 °C (1013 hPa) Melting Pt: 70 °C (dec.) Density: 1,998 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011331 CAS Number: 7783-00-8 EINECS: 231-974-7 UN: 3283 ADR: 6.1,II Merck Index: 14,08430 |
Specification Test Results
Appearance | White to off-white to light pink Crystalline powder or crystals and/or chunks |
Titration Iodometric | ≥97.5 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...