Sodium selenate 98%
Supplier: Thermo Scientific
|
Danger
|
369960250
369961000
369965000
ACRO369960250EA
63
EUR
ACRO369960250
ACRO369961000
ACRO369965000
Sodium selenate 98%
Sodium selenate
Formula:
Na₂O₄Se MW: 188,94 g/mol Density: 3,098 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003490 CAS Number: 13410-01-0 EINECS: 236-501-8 UN: 2630 ADR: 6.1,I Merck Index: 12,08820 |
Specification Test Results
Appearance | White Crystalline powder |
Assay | ≥97.5 % |
Identification | confirmed |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...