Potassium tellurite

Supplier: SOUTHERN GROUP
Danger

Synonyms: Potassium telluritePotassium tellurium trioxidePotassium tellurium (IV) oxide

SGRO0577EA 62.1 EUR
SGRO0577
Potassium tellurite
di-Potassium tellurium trioxide
Formula: K₂TeO₃
MW: 253,79 g/mol
Melting Pt: 460…470 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011379
CAS Number: 7790-58-1
EINECS: 232-213-1
UN: 3284
ADR: 6.1,II
Merck Index: 12,07850

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR