Sodium tellurite(IV) ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium tellurite, Disodium trioxotellurate

A15633.18 A15633.30
A15633.18EA 197 EUR
A15633.18 A15633.30
Sodium tellurite(IV) ≥97%
Sodium tellurite(IV)

Purity: 97% CAS No.: 10102-20-2 Molecular Formula: Na2O3Te Formula Weight: 221.58 Merck Reference: 14,8685 UN No.: 3284 MDL No.: MFCD00036141

Formula: Na₂O₃Te
MW: 221.58 g/mol
Melting Pt: 710
Storage Temperature: Ambient
MDL Number: MFCD00036141
CAS Number: 10102-20-2
EINECS: 233-268-4
UN: 3284
ADR: 6.1 II
Merck Index: 12,08834

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR