3-Quinuclidinone hydrochloride 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one hydrochloride

132260250 132261000 132265000
ACRO132260250EA 93.8 EUR
ACRO132260250 ACRO132261000 ACRO132265000
3-Quinuclidinone hydrochloride 99%
3-Quinuclidinone hydrochloride
Formula: C₇H₁₂ClNO
MW: 161.63 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00137391
CAS Number: 1193-65-3
EINECS: 214-776-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 % (On dry substance)
Loss on drying ≤1.0 % (100°C, 2 h)
Heavy metals ≤10 ppm
Sulfated ash ≤0.1 % (800°C)
UV at 280 nm A: ≤0.06
pH 6.0 to 9.5 (20°C, 10% in H2O)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.05 %
Ammonium (NH4) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR