Succinyl chloride ca. 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
132840250
132841000
132845000
ACRO132840250EA
42.5
EUR
ACRO132840250
ACRO132841000
ACRO132845000
Succinyl chloride ca. 95%
Succinyl chloride
Formula:
C₄H₄Cl₂O₂ Boiling Pt: 190 °C (760 mmHg) Melting Pt: 16…17 °C Density: 1,407 g/cm³ Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000749 CAS Number: 543-20-4 EINECS: 208-838-0 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4670 to 1.4720 (20°C, 589 nm) |
Titration RCOCl | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...