trans-Benzylideneacetone 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(E)-4-Phenylbut-3-en-2-one, trans-4-Phenyl-3-buten-2-one
164525000
164520500
164521000
ACRO164525000EA
328
EUR
ACRO164525000
ACRO164520500
ACRO164521000
trans-Benzylideneacetone 99%
trans-Benzylideneacetone
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Boiling Pt: 260…262 °C (760 mmHg) Melting Pt: 38.5…41 °C Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008779 CAS Number: 1896-62-4 EINECS: 217-587-6 |
Specification Test Results
Appearance | White to yellow Powder or crystals or solid |
Infrared spectrum | Conforms |
Melting point | 38.5 to 41 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...