(R)-(-)-β-Aminophenethyl alcohol 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

183820050 183820250
ACRO183820050EA 112 EUR
ACRO183820050 ACRO183820250
(R)-(-)-β-Aminophenethyl alcohol 98%
(R)-(-)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 79 °C
HPLC ≥97.5 %
Specific optical rotation -28° to -33° (20 °C, 589 nm) (C=1, 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR