(R)-(+)-Epoxystyrene 95%, 95% ee
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
(R)-(+)-Styrene oxide, (R)-Styrene oxide
240550050
240550010
240550250
ACRO240550050EA
120
EUR
ACRO240550050
ACRO240550010
ACRO240550250
(R)-(+)-Epoxystyrene 95%, 95% ee
(R)-(+)-Epoxystyrene
Formula:
C₈H₈O Boiling Pt: 192…195 °C (760 mmHg) Melting Pt: –37 °C Density: 1,053 g/cm³ Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00066210 CAS Number: 20780-53-4 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5340 to 1.5380 (20°C, 589 nm) |
GC | ≥94 % |
Enantiomeric excess | ≥94 % |
Specific optical rotation | +31° to +35° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...