(±)-β-Aminophenethyl alcohol 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
374280010
374280050
ACRO374280010EA
36.7
EUR
ACRO374280010
ACRO374280050
(±)-β-Aminophenethyl alcohol 98%
(±)-β-Aminophenethyl alcohol
Formula:
C₈H₁₁NO Boiling Pt: 261 °C (760 mmHg) Melting Pt: 53…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130145 CAS Number: 7568-92-5 UN: 3263 ADR: 8,III |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...