2,5-Dichloro-3-nitropyridine 97%

Supplier: Thermo Scientific
Warning

379520010 379520050
ACRO379520010EA 34.5 EUR
ACRO379520010 ACRO379520050
2,5-Dichloro-3-nitropyridine 97%
2,5-Dichloro-3-nitropyridine
Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 39…45 °C
Storage Temperature: Ambient
MDL Number: MFCD06658963
CAS Number: 21427-62-3

Order Now

Specification Test Results

Appearance Green-beige to orange Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 39 to 45 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR