4-Acetamidobenzaldehyde 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
400020100
400020250
400021000
ACRO400020100EA
86.9
EUR
ACRO400020100
ACRO400020250
ACRO400021000
4-Acetamidobenzaldehyde 98%
4-Acetamidobenzaldehyde
Formula:
C₉H₉NO₂ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003380 CAS Number: 122-85-0 EINECS: 204-579-2 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 154 to 158 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...