Bis(cyclopentadienyl)tungsten dichloride ≥99%

Supplier: Thermo Scientific
Danger


Packaged under argon.

40521.03 40521.MD
40521.03EA 351 EUR
40521.03 40521.MD
Bis(cyclopentadienyl)tungsten dichloride ≥99%
Bis(cyclopentadienyl)tungsten dichloride

Purity: 99% CAS No.: 12184-26-8 Molecular Formula: C10H10Cl2W Formula Weight: 384.95 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive UN No.: 3261 MDL No.: MFCD00269852

Formula: C₁₀H₁₀Cl₂W
MW: 384,93 g/mol
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00269852
CAS Number: 12184-26-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR