Cadmium(II) acetylacetonate ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Cadmium 2,4-pentanedionate, Cadmium acetylacetonate

41968.06 41968.14
41968.06EA 62.6 EUR
41968.06 41968.14
Cadmium(II) acetylacetonate ≥98%
Cadmium (II) acetylacetonate

Purity: 98% CAS No.: 14689-45-3 Molecular Formula: C10H14CdO4 Formula Weight: 310.64 Physical Form: Powder Melting Point: dec. UN No.: 2570 MDL No.: MFCD00040412

Formula: C₁₀H₁₄CdO₄
MW: 310,63 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00040412
CAS Number: 14689-45-3
EINECS: 238-730-9
UN: 2570
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR