Bis(cyclopentadienyl)zirconium dihydride ≥98%
Supplier: Thermo Scientific
|
Danger
|
44377.03
44377.06
44377.03EA
77.5
EUR
44377.03
44377.06
Bis(cyclopentadienyl)zirconium dihydride ≥98%
Bis(cyclopentadienyl)zirconium dihydride
Purity: 98%
CAS No.: 37342-98-6
Molecular Formula: C10H12Zr
Formula Weight: 223.42
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White
UN No.: 3131
MDL No.: MFCD04126436
Hydrogenation catalyst for alkenes and alkynes
Formula:
C₁₀H₁₂Zr MW: 223,43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04126436 CAS Number: 37342-98-6 UN: 3131 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...