Methyl-6-chloronicotinate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 6-chloronicotinate
A10396.06
A10396.14
A10396.06EA
55.8
EUR
A10396.06
A10396.14
Methyl-6-chloronicotinate ≥98%
Methyl 6-chloronicotinate
Purity: 98%
CAS No.: 73781-91-6
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Melting Point: 83-88°
Beilstein Reference: 128648
Formula:
C₇H₆ClNO₂ MW: 171,58 g/mol Melting Pt: 83…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023420 CAS Number: 73781-91-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...