Isatoic anhydride ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1H-Benzo[d][1,3]oxazine-2,4-dione

A14603.0E A14603.22 A14603.36
A14603.0EEA 321 EUR
A14603.0E A14603.22 A14603.36
Isatoic anhydride ≥97%
Isatoic anhydride

Purity: 97% CAS No.: 118-48-9 Molecular Formula: C8H5NO3 Formula Weight: 163.13 Flash Point: 308°(586°F) Melting Point: ca 237° dec. Beilstein Reference: 136786 MDL No.: MFCD00006700

Formula: C₈H₅NO₃
Melting Pt: ∼ 237 °C (dec.)
Density: 1,520
Flash Pt: 308 °C (586 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006700
CAS Number: 118-48-9
EINECS: 204-255-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR