D-(-)-α-Phenylglycine ≥99%

Supplier: Thermo Scientific

Synonyms: D-(-)-2-Phenylglycine, H-D-Phg-OH, (R)-(-)-α-Aminophenylacetic acid

A15669.14 A15669.22 A15669.36
A15669.14EA 31 EUR
A15669.14 A15669.22 A15669.36
D-(-)-α-Phenylglycine ≥99%
D(-)-α-Phenylglycine

Purity: 99% CAS No.: 875-74-1 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 277° dec. Beilstein Reference: 2208676 Merck Reference: 14,7291 Optical Rotation: -154° (c=1 in 1N HCl) MDL No.: MFCD00008061

Formula: C₈H₉NO₂
MW: 151,16 g/mol
Melting Pt: 277 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008061
CAS Number: 875-74-1
EINECS: 212-876-3
Merck Index: 14,7291

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR