Succinyl chloride 96%

Supplier: Thermo Scientific
Danger

Synonyms: Succinyl dichloride

A18389.18 A18389.30
A18389.18EA 75.5 EUR
A18389.18 A18389.30
Succinyl chloride 96%
Succinyl chloride

Purity: 96% CAS No.: 543-20-4 Molecular Formula: ClC(O)CH2CH2C(O)Cl Formula Weight: 154.98 Flash Point: 76°(168°F) Refractive Index: 1.4710 Odour: Acrid Melting Point: 16-17° Boiling Point: 190° Beilstein Reference: 507064 Merck Reference: 14,8873 UN No.: 3265 MDL No.: MFCD00000749

Formula: C₄H₄Cl₂O₂
MW: 154,98 g/mol
Boiling Pt: 190 °C
Melting Pt: 16…17 °C
Density: 1,407
Flash Pt: 76 °C (168 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000749
CAS Number: 543-20-4
EINECS: 208-838-0
UN: 3265
ADR: 8,II
Merck Index: 14,8873

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR