(R)-(+)-Epoxystyrene ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 20780-53-4
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 74°(165°F)
Refractive Index: 1.5350
Boiling Point: 194°
Beilstein Reference: 1984
Optical Rotation: +32° (neat)
UN No.: 2810
MDL No.: MFCD00066210
Formula:
C₈H₈O Boiling Pt: 194 °C Density: 1,051 Flash Pt: 74 °C (165 °F) |
MDL Number:
MFCD00066210 CAS Number: 20780-53-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...