5-Chloro-3-pyridineboronic acid 95%

Supplier: Thermo Scientific
Warning

H27756.06 H27756.03
H27756.06EA 0 EUR
H27756.06 H27756.03
5-Chloro-3-pyridineboronic acid 95%
5-Chloro-3-pyridineboronic acid

Purity: 95% CAS No.: 872041-85-5 Molecular Formula: C5H5BClNO2 Formula Weight: 157.37 MDL No.: MFCD06798243

Formula: C₅H₅BClNO₂
MW: 157,36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR