3-Aminocrotononitrile mixture E/Z 96%
Supplier: Thermo Fisher Scientific
L05316.22
L05316.36
L05316.22EA
48.5
EUR
L05316.22
L05316.36
3-Aminocrotononitrile mixture E/Z 96%
3-Aminocrotononitrile (mixture of cis and trans isomers)
Purity: 96%
CAS No.: 1118-61-2
Molecular Formula: C4H6N2
Formula Weight: 82.11
Storage Temperature: 2°C to 8°C
Flash Point: 154°(309°F)
Melting Point: ca 70-75°
Boiling Point: 120°/4mm
Beilstein Reference: 1719815
MDL No.: MFCD00008071
Formula:
C₄H₆N₂ Boiling Pt: 120 °C (4 mmHg) Melting Pt: ∼ 70…75 °C Flash Pt: 154 °C (309 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008071 CAS Number: 1118-61-2 EINECS: 214-266-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...